Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzonitrile, 98%

Catalog No. AAA1702914
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 368-77-4 | C8H4F3N | 171.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-77-4
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001807
InChI Key OGOBINRVCUWLGN-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzonitrile, 3-cyanobenzotrifluoride, m-trifluoromethylbenzonitrile, alpha,alpha,alpha-trifluoro-m-tolunitrile, benzonitrile, 3-trifluoromethyl, a,a,a-trifluoro-m-tolunitrile, unii-y7zd7dk3ay, alpha,alpha,alpha-trifluoro-m-toluonitrile, y7zd7dk3ay, a,a,a-trifluor-m-tolunitril
PubChem CID 67783
IUPAC Name 3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=CC(=C1)C(F)(F)F)C#N

Specifications

Melting Point 14°C to 18°C
Density 1.28
Boiling Point 188°C to 190°C
Flash Point 72°C (161°F)
Refractive Index 1.459
Quantity 25 g
UN Number UN3276
Beilstein 1868102
Formula Weight 171.12
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzonitrile
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 206-711-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.