Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Undecanol 94.0+%, TCI America™
SDP

Catalog No. U002825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U002825ML 25 mL
U00285ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U002825ML Supplier TCI America Supplier No. U002825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6929-08-4
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00046730
InChI Key HCARCYFXWDRVBZ-UHFFFAOYNA-N
PubChem CID 98970
IUPAC Name undecan-3-ol
SMILES CCCCCCCCC(CC)O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 mL
UN Number 1987
Formula Weight 172.31
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Undecanol
EINECSNumber (2)-0217&(2)-3704
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.