Learn More
VisionMate™ HSX High Speed Barcode Reader
GREENER_CHOICE
Catalog No. 312850
This item is not returnable. View return policy
Catalog No. 31-285-0 Supplier Thermo Scientific Supplier No. 312850
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The Thermo Scientific™ VisionMate™ HSX High Speed Barcode Reader decodes 2D tube barcodes in as little as one second. Compatible with Matrix™ , Nunc™ , and other brand tubes in 24, 48, 96, 138 (high density) and 384 format racks.
The Thermo Scientific™ VisionMate™ HSX High Speed Barcode Reader decodes 2D tube barcodes in as little as one second. It is compatible with Matrix™ , Nunc™, and other brand tubes in 24, 48, 96, 138 (high density) and 384 format racks. The redesigned VisionMate™ Suite software offers a range of export options for seamless transfer of barcode data to your LIMS, ELN, or database. Pre-loaded rack configurations allow rapid reading of 2D barcoded tubes without additional setup steps. The VisionMate™ HSX can decode both 2D and linear rack barcodes, providing versatile tracking options. Customizable LED visual feedback instantly notifies users of successful reads and any read errors. With an IP66 rating, the VisionMate™ HSX is protected from various adverse environmental conditions in your laboratory and makes it suitable for clean room use. The base unit is fully powered by a single USB connection to your computer. The VisionMate™ HSX can be used on the benchtop or integrated into your automated system.
  • Reads 2D barcoded Matrix™, Nunc™, and other brand tubes in 24, 48, 96, 138 (high density) and 384 format racks in as little as one second.
  • Graphical user interface provides multiple options for data viewing and export to your LIMS, ELN, or database.
  • Pre-loaded rack configurations for Matrix™ and Nunc™ racks.
  • Can read both 2D and linear rack barcodes for additional tracking options.
  • The included magnetic linear barcode reader can be positioned in various locations on the all metal housing of the base unit to read linear rack barcodes.
  • Cyroprotection provides resistance to the accumulation of frost on the reading surface.
  • Customizable LED feedback gives the user instant notification of successful reads and read errors relating to number of tubes, rack orientation, and rack barcodes.
  • Base reader is fully powered by a single USB connection to your computer.
  • IP66 Rating provides a critical environment barrier that enables clean room use and protects the reader from adverse environmental conditions.
  • Can be used as a benchtop unit or integrated into your automated system.
  • A perfect complement to the Thermo Scientific™ Matrix™ and Nunc™ 2D barcoded tube offerings, the VisionMate™ HSX provides valuable process flexibility and compatibility in storage workflows in drug discovery, biological, or genomic storage environments.
  • The VisionMate™ HSX is a valuable addition to storage processes which include other Thermo Scientific™ products:
    • Thermo Scientific™ Capit-All™ Flex Automated Decapper
    • Thermo Scientific™ 8-Channel Handheld Screw Cap Decapper
    •  
TRUSTED_SUSTAINABILITY
Specifications
Battery Run Time No battery, USB Powered
Decoding Capacity Single Tubes, Single Rack of Tubes (24, 48, 96, 138, and 384 Format Racks)
Data Transfer Method CSV, XLS, XML, Image, ODBC, TCP/IP
Includes Magnetic linear barcode reader
Dimensions (L x W x H) 7.9 x 5.9 x 6.3 in. (200 x 150 x 160 mm)
Warranty 12 months
Speed As little as 1 second per rack
Quantity Case of 1
Type High Speed Barcode Reader
Connection Type USB 2.0+
Data Memory None
Display Type None Included
Power Supply USB
Resolution No Display Included