Learn More
United Scientific Supplies 32 Ounce LDPE Bottles with Pump
SDPSCI_ED
Catalog No. S82817 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
No. per Pack:
12 Pk.
96 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
S82817 96 Pk.
S82816 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S82817 Supplier United Scientific Supplies Supplier No. UNSBLDPE32OZBULK
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
32 Ounce LDPE Bottles with Pump are made from LDPE and are useful for many applications.
  • Ideal for dispensing liquids such as hand sanitizer, soaps and more.
  • It is lightweight and has excellent durability.
  • 30 oz. capacity.
  • Available in pack and case quantities.
Specifications
Capacity (Metric) 946 mL
Capacity (English) 32 oz.
Color Opaque
Material LDPE
No. per Pack 96 Pk.
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Threaded
Shape Cylindrical
Product Type Pump Dispenser
Closure Material Plastic
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable