Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 36 in. Interior Shelf Kit
SDP

Catalog No. 01670278 Shop All Labconco Corporation Products
Change view
Click to view available options
Color:
White
Material:
Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
01-670-278 White Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-278 Supplier Labconco™ Supplier No. 9919101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

36 in. Interior Shelf Kit to fit within Protector Standard Storage Cabinet

  • These shelf kits fit within Protector Standard Storage Cabinets to provide additional surfaces to hold supplies and small laboratory instruments.
  • 36 in.

Specifications

Product Type Cabinet
Color White
Material Steel
For Use With (Equipment) Protector Standard Storage Cabinets
No. of Shelves 1
Width (English) 36 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.