Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson Gloves 360 Degree Nitrile 4 mil Gloves

Catalog No. 17099786
Encompass
Click to view available options
Size:
X-Large
2X-Large

Powder free 4 mil blue Nitrile Gloves offer superior strength with excellent resistance to punctures and tears, chemically resistant to oils, solvents and grease. Rolled cuff prevents liquid from getting inside the glove and adds extra strength to the cuff. Electrostatic discharge compliant.

  • Blue nitrile
  • Powder-free
  • 4 mil thickness
  • Rolled cuff
  • CFIA approved
  • 100 gloves/dispenser
  • Industrial grade; not for medical applications

Specifications

Certifications/Compliance CFIA
Certifications/Compliance (Europe) Nill
Accelerator Free No
Applications Industrial
AQL 4
Color Blue
Disposable Single-use
Exterior Finish Smooth
Finger Thickness (English) 4 mil
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4 mil
Powdered Powder Free
Product Type Nitrile Gloves
Size X-Large
Sterility Non-sterile
Thickness (English) 4 Mil
Length (English) 9 in.
Length (Metric) 22.85 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.