Learn More
ML Kishigo 3700 Series Incident Command Vest
Catalog No. 19441090
Click to view available options
Color:
Black
Dark Blue
Fluorescent Orange
Fluorescent Yellow
Green
Light Blue
Orange
Red
Royal Blue
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
ML Kishigo 3700 Series Non-ANSI Compliant Incident Command Vest
  • 100% polyester fabric with matching color trim
  • Hook and loop front closure
  • White 1 in. wide reflective material (White vest uses lime reflective material)
  • Front, clear card pocket holds 3x5 in. card
  • Back, clear card pocket holds 4.25x11 in. card
Specifications
Construction Solid Polyester
Certifications/Compliance Non-ANSI
Color White
Size One Size
Material Polyester
Length (Metric) 61 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.