Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 380 Foam Nitrile Coated Seamless Knit Nylon Gloves

Catalog No. 19021011 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-021-011 Medium
19-021-010 Small
19-021-012 Large
19-021-013 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-021-011 Supplier SHOWA™ Supplier No. 380M07
Add to Cart
Add to Cart
This item is not returnable. View return policy

General purpose gloves provide a long lasting grip in wet or dry conditions while also ensuring tactile precision while working with oily parts or lifting heavy objects.

  • Long-lasting grip in both wet and dry environments
  • Ensures tactile precision while working with small oily components or lifting heavy objects
  • Elasticated wrist cuff prevents debris from entering the glove
  • Open-back design allows air and moisture to escape

Specifications

Construction Seamless Knit
Coating Material Nitrile Foam
Color Blue
Hand Specific
Latex Free No
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
For Use With (Application) Automotive Repairs, and Intricate/Oily Parts Handling
Glove Cuff Style Knit
Series 380
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.