Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 381 Ultra-Lightweight Microfiber Gloves with Microporous Nitrile Coating

Catalog No. 19395345 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-395-345 X-Large
19-395-342 Small
19-395-343 Medium
19-395-344 Large
19-395-346 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-395-345 Supplier SHOWA™ Supplier No. 381XL09
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are designed with flexibility and comfort in mind, weighing only 19 grams with a finger thickness of 0.84 mm to ensure optimal dexterity when working on delicate projects.

  • Ultra-lightweight gloves designed with flexibility and comfort in mind
  • Microfiber construction with embossed microporous nitrile coating
  • Thin material ensures optimum tactility when working on delicate tasks
  • Breathable microfiber helps reduce palm sweat in warm environments
  • Nitrile coating enhances grip capabilities

Specifications

Construction Seamless Knit
Coating Material Microporous Nitrile
Color Gray, Black
Disposable Reusable
Finger Thickness (Metric) 0.84 mm
Hand Specific
Latex Free Yes
Material Microfiber
Product Type Seamless Knit Gloves
Size X-Large
For Use With (Application) Automotive Repairs, Component Assembly, Heavy Equipment Operation, Intricate Parts Handling, and Tool Operation
Glove Cuff Style Knit
Series 381
Yarn Weight 19 g
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.