Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 382 Microporous Nitrile Coated Nylon Gloves

Catalog No. 19021016 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-021-016 Large
19-021-014 Small
19-021-015 Medium
19-021-017 X-Large
19-021-018 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-021-016 Supplier SHOWA™ Supplier No. 382L08
Add to Cart
Add to Cart
This item is not returnable. View return policy

General purpose gloves designed to prevent fingerprints on metal sheets and glass. Nitrile coating disperses oil away to enhance grip for precision handling in greasy environments.

  • Gloves help to prevent fingerprints on metal sheets and glass
  • Microporous nitrile coating disperses oil away to enhance grip in greasy environments
  • Ergonomic design ensures comfort and breathability
  • Silicone-free

Specifications

Coating Material Microporous Nitrile
Color Blue, Black
Hand Specific
Latex Free Yes
Material Nylon
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Automotive Repairs, and Intricate/Oily Parts Handling
Glove Cuff Style Knit
Series 382

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.