Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin Hydrate 90.0+%, TCI America™
SDP

Catalog No. A21221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21221G 1 g
A2122200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21221G Supplier TCI America Supplier No. A21221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121916-94-7
Molecular Formula C36H61NO29
Molecular Weight (g/mol) 971.862
InChI Key TXBHONGFOHSVMN-IIRXRYJYSA-N
PubChem CID 131869226
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)N)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 971.86
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.