Learn More
3B Scientific™ 3B Birthing Simulator - Basic
GREENER_CHOICESDPSCI_ED
Catalog No. s27314 Shop All 3B Scientific Products
Click to view available options
:
basic birthing simulator
3B Scientific™ Birthing Simulator P90 BASIC has been developed for the skill training in normal deliveries, complicated deliveries and obstetric emergencies. Obstetric simulation has proven successful to enhance the training of delivery skills, following of protocols and reaction in emergency situation.
Simulator allows for assessment and manipulation of fetal positions as birthing complications are in general less likely when an abnormal fetal position or presentation can be detected before the labor process starts. When a fetus doesn't present in a normal position (facing backwards, presenting head first), the correct measures must be applied to assist a complicated delivery with manual maneuvers. Furthermore, medical staff must be trained on when and how to apply obstetric emergency interventions (like a cesarean section).
Users can see how manual maneuvers affect the fetus in-utero and train the following delivery skills and techniques:
  • Normal vaginal delivery
  • Normal delivery of umbilical cord and placenta
  • Palpation and assessment of fetal position and presentation
  • Diagnosis of all normal and abnormal fetal positions and presentations (like face, neck, or brow) to represent routine and difficult deliveries
  • Shoulder dystocia demonstration and management
  • Breech deliveries (including complete, footling or incomplete, and frank breech presentation)
  • Palpation of fetal fontanelles features
  • Complete and incomplete placenta birth
  • Demonstration of placenta previa: total, partial, and marginal
  • Prolapse of umbilical cord
  • Nuchal umbilical cord
  • Cesarean delivery through pre-cut incision
  • Clamping and cutting of umbilical cord
  • Simulated suctioning of baby's mouth and nose
  • Pinard's maneuver
  • Mauriceu-Smellie-Veit maneuver
  • Prague maneuver
  • Woods maneuver
  • Rubin
Offers the following features for effective obstetric teaching and training:
  • Fully articulating fetus with open mouth, tiltable head, shoulder blade, and anatomical correct fontanelles
  • Life-size adult female lower torso, anatomically correct landmarks like pelvis, pelvic cavity, and spine
  • Manual birthing system to control rotation and speed of fetus delivery
  • Easy to change position of fetus during birthing process including descend, flexion, extension, internal and external rotation, and restitution
  • Easy change-out of parts through pin connections, no tools necessary
  • Anti-slip base and handles for extra support on tables
Includes:
  • Birthing Simulator P90 BASIC
  • Detachable, padded stomach cover with basic insert with pre-cut caesarian section
  • Basic birth canal
  • Lubricant
  • Assembly kit
  • Fully articulating fetal baby
  • Umbilical cord
  • Placenta
Features:
  • Made of soft vinyl (hard for inner structures) and silicone.
  • Easy to clean
  • All parts can quickly be changed without tools (pin connections)
  • Can be upgraded to the PRO version with the Upgrade Kit 1020337.
Specifications
Product Type Birthing Simulator
For Use With (Application) Birthing Training
Includes Birthing Simulator, Detachable, stomach cover with basic insert and pre-cut caesarian section, Basic birth canal, Lubricant, Assembly kit, Fully articulating fetal baby, Umbilical cord, Placenta
Material Vinyl, Silicone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.