Learn More
3B Scientific™ Advanced Four-Vein Venipuncture Training Aid™ — Dermalike II™ Latex Free
SDPSCI_ED
Catalog No. s07960 Shop All 3B Scientific Products
Click to view available options
Product Type:
Dark Pigmentation
Light Pigmentation
Model permits the injection of fluids and withdrawal of blood.
Allows user to improve competency and technique by progressing from easier, larger veins to the more difficult, smaller and deeper veins
  • Four barely discernable blue veins in three different sizes, with the fourth vein deeply placed in a semi-transparent, tissue-like material
  • Pop is felt as the needle enters the vein, followed by a realisticflashbackof simulated blood, confirming the proper needle placement
  • Stain-resistant Dermalike™ material offers 50% less needle drag when accessing the underlying veins, improved tear resistance to allow twice as manysticks,nooily/stickysurface and requires no baby powder
  • The blue veins are made of Dermalike V™, a material with properties more similar to real veins, as you do not use excessive force to access the veins compared to the latex tubing still being used by other manufacturers
  • Size: 5 x 10 x 1.5 in.
Order Info
Carrying case (S07961) sold separately.
Specifications
Product Type Dark Pigmentation
For Use With (Application) Phlebotomy Training
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.