Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Basic Patient Care Manikin Upgrade Kit
SDPSCI_ED

Catalog No. S96633
Click to view available options
:
Upgrade Kit for P11 Patient Care Manikin
This item is not returnable. View return policy
This item is not returnable. View return policy

Allows practice of most training applications from basic to elderly patient care. Upgrade Kit includes internal organs for more advanced learning and teaching opportunities.

This upgrade kit easily converts a P11/1 Patient Care Manikin Basic to a P10/1 Pro version. Save money and upgrade your patient care manikin for more learning and teaching opportunities!

The kit includes:

  • Lung
  • Bladder
  • Stomach
  • Heart
  • Intestine
  • Genital, male and female

Specifications

Product Type Upgrade Kit for P11 Patient Care Manikin (Cat. No. S96632)
For Use With (Application) Allows Practice of Most Training Applications from Basic to Elderly Patient Care
Color Caucasian
Includes Lung, Bladder, Stomach, Heart, Intestine, Male and female genital organs (internal and external)
Material Plastic
Weight (English) 7.72 lb.
Weight (Metric) 3.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.