Learn More
3B Scientific™ Digital Teslameter with Probe
Click to view available options
:
Digital teslameter with probe
Description
- Includes a Hall sensor probe for measuring axial and tangential magnetic fields
- Probe also serves as a ruler as it includes a metric scale
- Teslameter can be calibrated by user
- Unit outputs a voltage proportional to the magnetic field which can be measured with a datalogger, XY-recorder or analog multimeter
Specifications
- Resolution: 0.01mT
- Digital Display: 3.5 digit LCD
- Input: BNC socket
- Output: 4mm safety sockets
- Dimensions of unit: 205 x 230 x 85 mm3
- Dimensions of probe: 360 x 15 x 25 mm3
Specifications
Specifications
Product Type | Digital Teslameter |
Includes | Hall sensor probe for measuring axial and tangential magnetic fields up to 200 mT |
For Use With (Application) | Allows students to incorporate quantitative measurements into their magnetism experiment |