Learn More
3B Scientific™ MDF™ Bravata Palm™ Aneroid Sphygmomanometer
SDPSCI_ED
Catalog No. s20680 Shop All 3B Scientific Products
Click to view available options
Size:
Adult
This item is intended for educational purposes only.
This item is intended for educational purposes only.
Includes
Nylon zippered carrying case with Velcro side pocket, ID tag.
Durable, accurate, and dependable blood pressure measurement device. The 3B Scientific MDF™ Bravata Palm™ Aneroid Sphygmomanometer has an extra-large gauge with high-contrast dial for easy and accurate readings.
  • Lightweight housing and bezel are constructed of high impact absorbing thermoplastic
  • Ergonomically-designed palm brace and bulb stabilize manometer and support hand during inflation and air release
  • Unsurpassed measurement, convenience and accuracy even in harsh field settings
  • Abrasion, chemical, and moisture resistant, adult Velcro™ cuff is constructed of high-molecular polymer nylon
  • Cuff is sized to meet American Heart Association recommendations
  • Artery indicator label and index range ensure proper cuffing for correct arterial compression
  • Single tube seamless inflation bag constructed of crack-resistant, non-sticking, high-density hypoallergenic latex-free PVC
  • Universal bladder tube enables compatible cuff exchange with all MDF and other major brands of BP systems
  • Chrome plated brass screw-type valve facilitates precise air release rate
  • Size: Adult
  • Color: Navy Blue
Warranty and Services
Three years and lifetime calibration.
Specifications
Size Adult
Includes Nylon zippered carrying case with Velcro side pocket, ID tag.
Latex Free Yes
Product Type Aneroid Sphygmomanometer
Color Navy Blue
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.