Learn More
3B Scientific™ ORTHObones Model of Mandible with Teeth
SDPSCI_ED
Catalog No. s07933 Shop All 3B Scientific Products
Click to view available options
:
Mandible with teeth
Resembles human bone in terms of form and characteristics.
  • Used by surgeons, orthopaedists and medical engineers worldwide
  • For demonstrating procedures like corrective jaw surgery or displaying medical devices used in craniomaxillofacial surgeries
  • Biomechanical realistic construction to simulate two distinct layers of real human bones; hard outer cortical shell and softer inner cancellous bone
Specifications
Dimensions (L x W x H) 4.3 x 3.9 x 2.4 in.
For Use With (Application) Demonstrating procedures like corrective jaw surgery; Displaying medical devices used in craniomaxillofacial surgeries
Weight (English) 0.15 lb.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.