Learn More
3B Scientific™ Set of 12 Materials with Four Different Weights of Each
SDPSCI_ED
Catalog No. s08160
Click to view available options
:
Twelve sets of test bodies made from same material but with four different weights
This item is not returnable. View return policy
This item is not returnable. View return policy
Twelve sets of test bodies to derive concept of density.
  • Made of same material but with four different weights
  • Materials: Wood, polypropylene, polyamide, acrylic (two colors), polyurethane, phenol, PVC (Three colors), PTFE and aluminum
  • For school experiments
  • Supplied in storage containers
Specifications
For Use With (Equipment) 100 mL graduated cylinder
Includes Wood, polypropylene, polyamide, acrylic (2 colors), polyurethane, phenol, PVC (3 colors), PTFE and aluminum; Supplied in storage containers
For Use With (Application) Deriving the concept of density in school experiments
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.