Learn More
3B Scientific™ Student Mechanics Experiment Set
SDPSCI_ED
Catalog No. s96474 Shop All 3B Scientific Products
Click to view available options
:
Mechanics experiment set
Includes
CD with experiment instructions; steel leaf spring; lever arm; ruler; pulley; pulley with hook; 100g weight; 50g weights (3); 25g weights (2); 2N dynamometer; metal pivot; washer rings to fit metal pivot (4); pulley with cord; trolley; friction pad; coil spring; iron block; aluminum block; wooden block; silicone tube, 500 x 3mm dia.; tubing connector; 8mm dia. clip; 14mm dia. clip; 50mL syringe; 30mL syringe; plastic sphere; rubber stopper, 30 x 31/25mm dia.; 40mm dia. funnel; U-tube manometer; measuring cylinder; and 100g modelling clay
Requires
A3B Student Experiment Basic Set
Set of apparatus for 26 basic experiments on the mechanics of solids, liquids, and gases
For use with 3B Scientific Student Experiment Basic Supply Set.
  • Packed in a robust plastic box with foam inserts moulded to the shapes of the items and a transparent lid
Specifications
Product Type Kit, Basic Mechanics
For Use With (Application) For 26 basic experiments on the mechanics of solids, liquids and gases
Includes CD with experiment instructions, steel leaf spring, lever arm, ruler, pulley, pulley with hook, 100g 3 weights, 50g 2 weights, 25g weights, 2N dynamometer, metal pivot, 4 washer rings to fit metal pivot, pulley with cord, trolley, friction pad, coil spring, iron block, aluminum block, wooden block, 500 x 3mm dia. silicone tube, tubing connector, 8mm dia. clip, 14mm dia. clip, 50mL syringe, 30mL syringe plastic sphere, 30 x 31/25mm dia. rubber stopper, 40mm dia. funnel, U-tube manometer, measuring cylinder and 100g modeling clay
Weight (English) 6.83 lb.
Weight (Metric) 3098 g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.