Learn More
3B Scientific™ Student Optics Experiments Supply Set
SDPSCI_ED
Catalog No. s96476 Shop All 3B Scientific Products
Click to view available options
:
Optics experiment set
Includes
CD with experiment instructions; screen; optical lamp; lens, f = 50mm; lenses, f = 100mm (2); lens, f = 300mm; lens, f = -100mm; slide-holder; semi-circular shape; lens shapes: rectangular, concave, and convex; prism; single-slit aperture; triple-slit aperture; aperture in the shape of an F; mirror with magnetic holder; small candles (2); opaque object; color slide; aluminum rod; overlay mask protractors (10); 2mm transparency film, DIN A5 size; and plug-in power supply unit for optical lamp
Requires
3B Scientific Student Experiment Basic Set
Set of apparatus for carrying out 19 basic student experiments in optics
For use with 3B Scientific Student Experiment Basic Set.
  • Packed in a robust plastic box with foam inserts moulded to the shapes of the items and a transparent lid
Specifications
Product Type Student Optics Experiments Supply Set
For Use With (Application) For carrying out 19 basic student experiments in optics
For Use With (Equipment) 3B Scientific student experiment basic set
Includes CD with experiment instructions, screen, optical lamp, lens, slide-holder, semi-circular shape, lens shapes, rectangular, concave and convex, prism, single-slit aperture, triple-slit aperture, aperture in the shape of an F, mirror with magnetic holder, 2
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.