Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ 3BP5L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558232
Description
Immunogen sequence: TQGSHLGSCI EKARPYYEAR RLAKEAQQET QKAALRYERA VSMHNAAREM VFVAEQGVMA DKNRLDPTWQ EMLNHATCK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
3BP5L | |
Polyclonal | |
Unconjugated | |
SH3BP5L | |
KIAA1720; SH3 binding domain protein 5 like; SH3 domain-binding protein 5-like; SH3-binding domain protein 5 like; SH3-binding domain protein 5-like; SH3BP5L; SH3BP-5-like; UNQ2766/PRO7133 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L8J4 | |
SH3BP5L | |
Recombinant protein corresponding to Human 3BP5L. Recombinant protein control fragment (Product #RP-102805). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction