Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 3BP5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558233 Supplier Invitrogen™ Supplier No. PA558233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQQAEARVQA LQKTLRRAIG KSRPYFELKA QFSQILEEHK AKVTELEQQV AQAKTRYSVA LRNLEQISEQ IHARRRGGLP PHPLGPRRSS PVGAEAGPED Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 3BP5L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3BP5L
Gene Accession No. Q7L8J4
Gene Alias KIAA1720; SH3 binding domain protein 5 like; SH3 domain-binding protein 5-like; SH3-binding domain protein 5 like; SH3-binding domain protein 5-like; SH3BP5L; SH3BP-5-like; UNQ2766/PRO7133
Gene Symbols SH3BP5L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human 3BP5L. Recombinant protein control fragment (Product #RP-94464).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.