Learn More
Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used, industry-standard cylindrical tanks. The tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
$285.00 - $447.00
Catalog Number | Mfr. No. | Capacity (Metric) | ForUseWithApplication | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | ForUseWithApplication | Price | Quantity & Availability | ||||
SH3098801
![]() |
Thermo Scientific™
SH30988.01 |
50 L |
Each for $285.00
|
|
|||||
SH3098802
![]() |
Thermo Scientific™
SH30988.02 |
100 L |
Each for $372.00
|
|
|||||
SH3098803
![]() |
Thermo Scientific™
SH30988.03 |
200 L |
Each for $447.00
|
|
|||||
Description
- Minimizes bioburden due to construction under sterile cGMP conditions
- Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
C-Flex |