Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ 3dGRO™ Human Colon Organoid Expansion Medium

Catalog No. scm304mi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM304MI 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM304MI Supplier MilliporeSigma™ Supplier No. SCM304
Only null left
Add to Cart
Add to Cart

Complete serum-free organoid media for expansion and 3-D culture of human intestinal colon organoids

  • Medium has been validated for use with 3dGRO™ Human iPSC Derived Colon Organoids
  • Colon organoids propagated in the medium express colon-specific markers including the posterior hindgut marker CDX2, a-carbonic anydrase II (CA-II), a-carbonic anhydrase IV (CA-IV), and goblet cell markers Mucin-2 and Mucin-5B
  • No growth/Pass
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt -20 °C, Ready to use 2 to 8 ° overnight thaw, Use immediately store (at 2°C to 8°C up to 1 week, Do not re-freeze)
Product Type Organoid Expansion Medium
Quantity 50 mL
Endotoxin Level <2 EU/mL
For Use With (Application) Cell Culture, Stem Cell Culture
Osmolality 350 to 375 mOsm
pH 7.0 to 7.4
Form Liquid

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.