Learn More
Epredia™ 3DHistech HistoQuant Image Segmentation Module - 3 Users
Shop All Epredia Products
Click to view available options
:
3 Users
Description
- Color- and pattern-based image analysis solutions are used to identify different tissue elements in stained tissue. Instead of the time-consuming manual annotation, automated detection and annotation of a region of interest is available with a quick training on relevant tissue samples.
- HistoQuant is an image segmentation module which identifies stained tissue elements based on color and intensity features. This is an adequate solution for double-stain quantification or multiplex fluorescent analysis, with dual stain colocalization option. This module can analyze fluorescent and brightfield slides.
- License for 3 users - Research use only
Specifications
Specifications
| Product Type | Image Segmentation Module |
| For Use With (Application) | Digital Pathology |
| For Use With (Equipment) | Microarray Scanner |
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.