Learn More
Epredia™ 3DHistech Pannoramic 1000 Digital Scanner, Brightfield Slide to 80x Resolution, Water Immersion
Catalog No. P100040808S Shop All Epredia Products
Change view
Click to view available options
80x
Catalog No.
P100040808S 80x
1 options
Catalog No. P100040808S Supplier Epredia™ Supplier No. P1000BF408080S
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic 1000 whole-slide digital scanner is specifically designed for high volume and high throughput labs that don't want to sacrifice flexibility and quality.
  • Largest-capacity, whole-slide scanner on the market: 1000 standard-width slides
  • Can scan 15 x 15 mm slide area in 30 seconds (20x objective (40x optical equivalent), 0.25 μm/pixel, single layer)
  • Touch-enabled, easy-to-use software interface
  • Double-width slide capacity (optional)
  • Automated tissue detection and coverslip detection
  • Flexible Priority/STAT slide position allocation
  • Impressive Speed
  • Multilayer (Z-stack) and Extended Focus scanning (optional)
  • 2 individual objectives exchangeable automatically during scanning
  • 1D and 2D barcode reading and parsing - Research use only
Specifications
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.