Learn More
Epredia™ 3DHistech SlideCenter Slide Management System - 50 Users
Shop All Epredia ProductsDescription
SlideCenter is a flexible, distributed server system: more servers can easily be added for enhanced data security and storage requirements.
Licensing is performed on the server side: SlideCenter regulates the number of users who can access the server at any time.
Five user types can be defined: Administrator user, Moderator user, Normal user, Restricted user and Public Share user.
Key Features
- Automatic slide alignment
- Virtual Slide and VirtualTray
- TMA workflow support
- Transparent login method for high-traffic user authentication
- Server-side barcode parsing
- InstantViewer
- Web-based slide and case database with fast search
- Easy extension by adding new SUs
- Slide access through the free SlideViewer, the InstantViewer or the iPad Viewer
- Virtual Slide handling
- MS Network, HTTP and HTTPS accessibility
- Fully manageable user and user group rights system
- Direct scanning into database with PANNORAMIC digital slide scanners
- Macro images and multiple file attachments stored in the case
- SlideCenter can work with other 3DHISTECH software applications (e.g. quantification software can be run on remote slides)
- License for 50 users - Research use only.
Specifications
Specifications
| Product Type | Slide Management System |
| For Use With (Application) | Digital Pathology |
| For Use With (Equipment) | Microarray Scanner |
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.