Learn More
Epredia™ 3DHistech TMA Master II Automated Tissue Microarrayer, 5 Block Capacity with PCR
Catalog No. TMA2PCR Shop All Epredia Products
Change view
Click to view available options
For Use With (Application):
Digital Pathology
Catalog No. For Use With (Application)
TMA2PCR Digital Pathology
1 options
Catalog No. TMA2PCR Supplier Epredia™ Supplier No. TMA2PCR
Epredia™ 3DHistech TMA Master II is the smallest fully automated tissue microarrayer on the market, it easily fits any laboratory bench.
  • TMA Master II is a robust and reliable computer-controlled instrument for creating tissue microarray (TMA) blocks.
  • It is an ideal solution for smaller laboratories, helping save reagent, slides, time, workforce and money.
  • By using TMA Master II, you can easily and quickly create TMA blocks with great precision, of 0.6, 1, 1.5 and 2 mm diameter tissue cores.
  • Optionally, the instrument can also extract tissue samples to standard 0.2 mL PCR tubes.
  • The extracted tissue samples can then be used later for various applications at the field of molecular pathology - Research use only.
Specifications
Product Type Automated Tissue Microarrayer
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.