Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 3G11 Sialoganglioside Antigen Monoclonal Antibody (eBio3G11 (SM3G11, 3G11)), eFluor™ 660, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 50544382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-544-382 100 μg
50-544-380 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-544-382 Supplier Invitrogen™ Supplier No. 50544382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eBio3G11 MAb reacts with the mouse 3G11antigen. The 3G11 antibody was isolated as an autoantibody against the 3G11 sialylated carbohydrate antigen, which was found to be expressed by thymoyctes and peripheral T cells, but not by B cells, myeloid cells or erythrocytes. 3G11 is expressed at higher levels on CD4+ T cells than on CD8+ T cells, however expression on both of these subsets decreases with advancing age. In thymus, 3G11 is expressed on approximately 20% of total thymocytes. 3G11 has also been used as a marker to distinguish naive and memory T cells, as 3G11 is irreversibly down-regulated upon activation of CD4+ T cells. The 3G11 antibody is able to block binding of antibodies to CD3 epsilon, suggesting close proximity of these two proteins. Recently, it has been demonstrated that loss of 3G11 expression in experimental autoimmune encephalomyelitis (EAE) is associated with an anergic, regulatory T phenotype. Purified 3G11- T cells were able to suppress proliferation and cytokine production of 3G11+ T cells, and part of this effect was due to the secretion of IL-10.

The 3G11 sialylated carbohydrate antigen was found to be expressed by thymoyctes and peripheral T cells, but not by B cells, myeloid cells or erythrocytes. 3G11 is expressed at higher levels on CD4+ T cells than on CD8+ T cells, however expression on both of these subsets decreases with advancing age. In thymus, 3G11 is expressed on approximately 20% of total thymocytes. 3G11 has also been used as a marker to distinguish naive and memory T cells, as 3G11 is irreversibly down-regulated upon activation of CD4+ T cells. 3G11 antibodies can be used to block binding of antibodies to CD3 epsilon, suggesting close proximity of these two proteins. Recently, it has been demonstrated that loss of 3G11 expression in experimental autoimmune encephalomyelitis (EAE) is associated with an anergic, regulatory T phenotype. Purified 3G11- T cells were able to suppress proliferation and cytokine production of 3G11+ T cells, and part of this effect was due to the secretion of IL-10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 3G11 Sialoganglioside Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone eBio3G11 (SM3G11, 3G11)
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias 3G11 antigen
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

Is the eFluor 660 fluorochrome compatible with Anti-Cy5/Alexa Fluor 647 beads?

Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.