Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 7800S Reusable Full Facepiece

Catalog No. 17663B Shop All 3M Products
Click to view available options
Product Type:
Full Facepiece
Full Facepiece Respirator
Format:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Compatible Series 2000 or 7093 filters or Series 6000 cartridges. (See separate listings.)

Can be used with 3M Series 2000 or 7093 filters or Series 6000 cartridges

When combined with appropriate attachments, these facepieces are NIOSH approved for use as air-purifying respirators, continuous-flow (Type C) airline respirators, or combination air-purifying and Type C respirators.

  • Soft, pliable facepiece made of easy-to-clean silicone with a double-flap seal
  • Extra-large inhalation and exhalation valves
  • Six adjustable straps to distribute weight
  • Standard nose cup to prevent fogging
  • Large faceshield with scratch-resistant polycarbonate lens
  • High-quality rust-free speaking diaphragm
  • 3M Catalog# 7800S-L , 3M Catalog# 7800S-M , 3M Catalog# 7800S-S

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Order Info

To convert to a Powered Air Purifying Respirator (PAPR) system, add 3M PAPR assembly and breathing tube. Order attachments separately on www.fishersafety.com.

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH 42CFR84
Connection Type Bayonet, DIN
Material Silicone, Polycarbonate Lens
Product Type Full Facepiece Respirator
Format Medium
For Use With (Application) Mining, oil and gas, construction, general manufacturing, transportation
For Use With (Equipment) 3M™ series 2000 or 7093 filter or series 6000 cartridge
Model 7800S-M
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.