Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ R95 Particulate Respirator, 8240
Shop All 3M Products
Click to view available options
:
R95 Particulate Respirator
Description
Economical R95 disposable particulate respirator features an adjustable nose clip and cushioning foam to provide a custom and secure seal against certain oil and non oil based particles.
- NIOSH approved R95 rating
- Adjustable nose clip
- Nose foam
- Braided headbands
Specifications
Specifications
| Aerosol Type | R - Somewhat Resistant to Oil |
| Assigned Protection Factor (APF) | 10 |
| Certifications (NIOSH) | NIOSH 42 CFR 84, R95, approval number: TC-84A-1350 |
| Certifications (OSHA) | OSHA 1910.134 |
| Color | White |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | R95 |
| Material | Polypropylene, Polyester |
| Product Type | General Purpose Particulate Respirator |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction