Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Powerflow™ High Efficiency SP3 Particulate Filter

Catalog No. 19019040 Shop All 3M Products
Change view
Click to view available options
:
SP3 Particulate filter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-019040 SP3 Particulate filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-019040 Supplier 3M™ Supplier No. 7000131701

Due to product restrictions, please to purchase or view availability for this product.

High Efficiency (HE) SP3 particulate filter system component is designed for use with Powerflow™ PAPR systems.

  • 3M Catalog# 024-00-02R01 , 3M Catalog# 450-01-01R20

Note: These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH
Color Magenta
Product Type Sp3 HEPA Filter
For Use With (Application) Ideal to protect against dusts, mists, fumes, asbestos, radionuclides and radon daughters
For Use With (Equipment) Powerflow™ PAPR Systems
Model 450-01-01R20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.