3M™ Disposable Protective Coverall 4530

Manufacturer: 3M™ 4530CSBLK4XL

 View more versions of this product

Catalog No. 17-624-120

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications/Compliance CE III, Type 5/6, Certified to European Standards to help protect against certain industrial hazards
    Material Breathable SMS
    Seam Type Serged and Bound
    Color Blue
    Size 4XL
    Coating Anti-static
    Disposable Yes
    Size (Metric) Chest 132 to 140cm
    Height (Metric) 200 to 206cm
    For Use With (Application) Construction, mining, oil and gas, Powder handling, abatement and remediation, cleaning and maintenance, provide basic barrier protection against light liquid splashes and hazardous dusts
    Latex Free No
    Collar Type Standard
    Closure Type Two-way zipper
    Size (English) Inseam 36 in.
    Size (English) Waist 55 in.
    Size (Metric) Waist 139cm
    Construction Suit: SMS Polypropylene, Back panel: SMS Polypropylene, Zipper: Nylon on Polyester Braid, Elastic: Neoprene Rubber, Cuffs: Polyester, Thread: Polyester
    Ankle Style Elastic
    Size (English) Chest 52 to 55 in.
    Height (English) 78 to 81 in.
    Cuff Style Knit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit