Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Coverall 4530

Catalog No. p-4702631 Shop All 3M Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Size (English) Chest:
36 to 39 in.
39 to 43 in.
43 to 45 in.
45 to 49 in.
49 to 52 in.
52 to 55 in.
Height (English):
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
78 to 81 in.
Size (English) Waist:
50 in.
51 in.
52 in.
53 in.
54 in.
55 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Disposable coveralls offer protection against certain light liquid splashes and hazardous dust. They feature a low-profile, three-panel hood that zips up to the chin for additional head protection.

  • Breathable material (SMMS) to help reduce heat build up
  • Elastic waist and ankles for convenience and freedom of movement
  • Knit cuffs for increased comfort
  • Standard collar provides convenience when using auxiliary PPE
  • Two-way zipper with sealable storm flap for convenience
  • Flame retardant coating offering limited protection against heat and flame
  • Reinforced gusset
  • Anti-static treatment on both sides
  • Protects against dusts and light liquid splashes
  • Typical applications may include welding, metal grinding, general powder handling, and woodworking
  • Designed to be worn over primary thermal protective FR garments
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Fabric
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged, Bound
Sterility Non-sterile
Coating Anti-static
Collar Type Standard Fold
Certifications/Compliance (Europe) EN 13034, EN 13982, EN 1149, EN 1073, ISO 14116
Description Suit: SMS Polypropylene, Back panel: SMS Polypropylene, Zipper: Nylon on Polyester Braid, Elastic: Neoprene Rubber, Cuffs: Polyester, Thread: Polyester
Latex Free No
Model 4530CS
Size (English) Chest 39 to 43 in.
Size (English) Inseam 32 in.
Size (English) Waist 51 in.
Size (Metric) Chest 100 to 108 cm
Size (Metric) Waist 129 cm
For Use With (Application) Construction, Mining, Oil and Gas, Abatement, Remediation, Cleaning, and Maintenance
Height (English) 69 to 71 in.
Height (Metric) 174 to 181 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.