Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Coveralls 4540+, Collared

Catalog No. 17624110 Shop All 3M Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls are designed to help protect against hazardous dusts (Type 5) and certain light liquid splashes (Type 6).

  • Premium dual material technology provides both protective performance and comfort
  • Barrier against dry particles and limited liquid chemical splash (CE Type 5/6)
  • Soft and light, super low-linting laminated microporous material
  • Back panel made of breathable SMS material for improved air circulation and reduced heat stress
  • Two-way zipper with fully sealable storm flap
  • Fold down collared design
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static treated
  • Low linting
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Polyethylene, Polyester/Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Bound
Sterility Non-sterile
Resistance Anti-static, Dust, Liquid
Coating Anti-static
Collar Type Standard Fold
Certifications/Compliance ANSI 101-1996
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Latex Free No
Model 4540+
Size (English) Chest 52 to 55 in.
Size (English) Inseam 36 in.
Size (English) Waist 55 in.
Size (Metric) Chest 132 to 140 cm
Size (Metric) Waist 139 cm
For Use With (Application) Pesticide, Asbestos and Lead Abatement, Power Plants, Metals, Cleaning, Maintenance, General Industrial, Insulation, Food Processing, Pharmaceutical
Height (English) 78 to 81 in.
Height (Metric) 200 to 206 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.