Learn More
3M™ H-700 Series Cap Style Vented Hard Hats
Catalog No. p-5211640 Shop All 3M Products
Click to view available options
Color:
Black
Blue
Green
Pink
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Helps protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension allows to customize height and fit
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Replaceable brow pad for comfort and extends service life
  • Short brim for upward visibility
  • Available in various colors
  • 3M Catalog# H-701V, H-702V, H-703V, H-704V, H-711V, H-712V, H-713V
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2014, Type 1
Color Blue
Includes Standard brow pad
Model H-703V
Product Type Cap-style Hard Hat
Quantity 20/Cs.
Suspension Type 4-Point Ratchet
Ventilated Ventilated
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.