Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Tychem™ Hoods and Head Covers

Catalog No. p-2360384 Shop All 3M Products
Change view
Click to view available options
Product Type:
Headcover
Respirator Hood
Size:
Large
Standard
Suspension Type:
Sewn-in Elastic Headband
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size Suspension Type
17-987-255B Respirator Hood Standard Sewn-in Elastic Headband
18-778B Headcover Large Sewn-in Elastic Headband
18-778A Headcover Standard Sewn-in Elastic Headband
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-987-255B Supplier 3M™ Supplier No. 7000005591

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Help protect against airborne particulates that have a concentration of up to 25 x PEL using head cover, or 1000 x PEL using hood

Hood (BE 10)

  • Includes disposable polycoated Tychem hood that covers the shoulders and chest, with a clear, wide-view acetate visor
  • Air enters from the breathing tube (sold separately), passes over the user's head and out at the base of the cape
  • Accommodates most facial hair and spectacles Weighs 3.2 oz. (0.1kg)
  • Supplied in packs of three hoods

Head Cover (BE 12)

  • Includes a liquid-repellent, polycoated Tychem head cover with a clear, wide-view acetate visor.
  • Comes with three regular or large-sized head covers in choice of white or blue

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.

  • 3M Catalog# BE-10-3, BE-12-3, BE-12-50, BE-12L-3
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA
Color White
Material Tychem QC
Product Type Respirator Hood
Size Standard
For Use With (Application) Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare
For Use With (Equipment) 3M™ air-mate™ and breathe easy™ powered air purifying respirator system and be series supplied air system
Includes Wide view acetate visor
Material (Lens) Acetate
Model BE-10-3
Suspension Type Sewn-in Elastic Headband
Weight (English) 3.2 oz.
Weight (Metric) 0.1 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.