Learn More
3M™ Maxim 2X2 Air Flow Goggles
Designed for tactical use, the Maxim 2x2 protective, polycarbonate air flow goggle is both stylish and durable. Featuring flexible fit temples, a vented anti-fog seal and an elastic strap for a secure fit, it maximizes visual comfort and clarity.
$76.00
Specifications
Lens Coating Anti-fog
View More Specs
Products 2
Catalog Number Mfr. No. LensType Price Quantity & Availability  
Catalog Number Mfr. No. LensType Price Quantity & Availability  
19-065-420
View Documents
3M™
7000127227
Each for $76.00
Case of 10 EA for $721.00
Only null left
Add to Cart
19-065-419
View Documents
3M™
7000127226
N/A N/A N/A
Description
Description
The Maxim 2X2 air flow safety goggle is a preferred solution for people in tactical situations. The lightweight safety goggle was designed to help protect from sun, wind, dust and flying debris, while incorporating expansive top and bottom venting to reduce fog and heat. Its dual-lens design enhances sightline and field of vision.
  • Anti-fog lens.
  • Nylon frame
  • Elastic strap
  • Meets the High Impact Requirements of ANSI Z87.1-2010
  • 10 safety goggles per case
  • Vented seals and anti-fog coating keeps your vision clear
  • Multi-protection goggles help protect from wind, dust, and flying debris
  • Choice of elastic strap for a secure fit or flexible fit temples
  • Available in two lens tints – clear and gray
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • 3M Catalog# 40698-00000 , 3M Catalog# 40699-00000
Specifications
Specifications
Anti-fog
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.