Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Nitro™ Ear Plugs

NRR: 32 dB

Specifications

Color Multi-color
Certifications/Compliance CSA Class AL
Size Regular
For Use With (Application) For easy insertion and canal-hugging seal
Noise Reduction Rating (NRR) 32 dB
View More Specs

Products 1
Catalog Number Mfr. No. Corded Price Quantity & Availability  
Catalog Number Mfr. No. Corded Price Quantity & Availability  
19-130-0465
View Documents
3M™
7000127250
Uncorded N/A N/A N/A
Description

Description

  • Ultra-soft foam formulation for all-day comfort
  • Tapered shape for easy insertion and canal-hugging seal
  • Four vibrant colors in each box: orange, red, pink, and purple
  • 3M Catalog# P1000 , 3M Catalog# P1001

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Multi-color
Regular
32 dB
Nitro™ Earplug
CSA Class AL
For easy insertion and canal-hugging seal
Tapered
Polyurethane
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.