Learn More
3M™ No-Touch™ Foam Ear Plugs P2000
Shop All 3M Products
Click to view available options
Model:
P2000
P2001
Corded:
Corded
Uncorded
Description
- Push-to-fit foam ear plugs create a hygienic, noise-reducing seal
- Design features soft foam, a smooth tapered shape, and paddle stems
- Soft purple foam is fused with the LiveWire stem
- Foam tips stay clean even if wearer's hands are dirty
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| ANSI S3.1974 | Yes |
| Cord Color | Yellow |
| Cord Material | Vinyl |
| CSA Z94.2 | Class AL |
| Size | Universal |
| For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
| High Visibility | Yes |
| Model | P2001 |
| Packaging Type | Poly bag |
| Pairs per Package | 400 Pairs/Cs |
| Show More |