Learn More
3M™ Nuvo™ Readers Eyewear
Shop All 3M Products
Click to view available options
Lens Tint:
Clear
Gray
Magnification Power:
+1.50
+2.00
+2.50
Description
Designed for workers who have difficulty reading small print or who are engaged in fine detail work.
- Classic look of a dual lens but with tough, lightweight one-piece lens
- Lens with anti-fog coating absorbs 99.9% UV
- Gray, adjustable length temples with soft tips for comfortable fit
- Soft nosepiece
- Integral sideshields provide profile protection
- Cushioned brow bar
- 3M Catalog# 11434-00000-20, 11435-00000-20, 11436-00000-20, 11500-00000-20, 11501-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Bifocal Readers |
| For Use With (Application) | Designed for workers who have difficulty reading small print or who are engaged in fine detail work |
| Magnification Power | +1.50 |
| Model | 11434-00000-20 |
| UV Protection | UV400 |