Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ CH-3 Headset with PTT

PELTOR™ CH-3 Headsets with an integrated Push-To-Talk (PTT) button on the ear cup allows for a direct connection to handheld communication devices.

$336.00 - $385.35

Specifications

Color Bright Yellow
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Manufacturing
High Visibility Yes
View More Specs

Products 3
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
19-072-241
Accessories View Documents
3M™
7100203638
MT74H52A-111 Over-the-head 26 dB
Each for $341.25
Case of 10 EA for $3,412.48
Only null left
Add to Cart
 
19-072-252
Accessories View Documents
3M™
7100205729
MT74H52B-111 Neckband 24 dB
Each for $385.35
Case of 10 EA for $3,853.51
Only null left
Add to Cart
 
19-072-300
Accessories View Documents
3M™
7100203150
MT74H52P3E-111 Hard Hat 24 dB
Each for $336.00
Case of 10 EA for $3,359.98
Only null left
Add to Cart
 
Description

Description

  • Protective communication solution for hands-free, clear communication in noisy environments
  • FLX2 connection jack for easy and reliable connection to a wide assortment of handheld devices
  • Designed to withstand harsh environmental elements
  • Water and dust-proof noise-canceling microphone (IP68) for clear speech transmission in noisy and harsh environments
  • Audio speakers are optimized for speech frequencies and meet the efficiency demands of many of today's digital devices
  • A Push-To-Talk button is conveniently located on the ear cup for easy access
  • Bright yellow color for enhanced visibility

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Bright Yellow
Black
Yes
CH-3 Headset with PTT
Yes
Manufacturing
1 Ea.
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.