3M™ Pistonz™ Push-in Foam Ear Plugs

NRR 29dB

Manufacturer: 3M™ P1401

 View more versions of this product

Catalog No. 19-177-951

  Check Availability
Add to Cart

Alert

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.


    Specifications

    Material Polyurethane
    Shape Push-to-fit Foam
    Certifications/Compliance CSA Class AL
    Product Type Pistonz™ Push-In Foam Earplug
    Hearing Protection Style Push-to-fit
    Style Corded
    Noise Reduction Range (NRR) 29dB
    Size One size fits to all
    Color Silver
    Cord Material Vinyl
    Dielectric Yes
    For Use With (Application) Metallic-style earplug tips with ultra-soft feel for all day wear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit