Learn More
3M™ Polycarbonate Faceshields
Protects workers from infrared hazard exposure.
$89.36 - $3097.50
Specifications
For Use With (Equipment) | 3M H-Series Hard Hats/Headgear |
---|---|
Lens Material | Polycarbonate |
Includes: Faceshield only. (Head gear is sold separately.)
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
19-040-047
|
3M™
7000127241 |
Shade 5 |
Case of 10 for $1,463.70
|
|
|||||
19-040-051
|
3M™
7000052753 |
Clear |
Each for $89.36
Case of 10 EA for $893.55
|
|
|||||
19-040-052
|
3M™
7010384061 |
Medium Green |
Case of 10 for $660.45
|
|
|||||
19-040-054
|
3M™
7000144939 |
Medium Green, Gold-plated |
Case of 10 for $3,097.50
|
|
|||||
19-040-055
|
3M™
7000127239 |
Gold-plated, Dark Green, Gold-plated |
Case of 10 for $3,034.50
|
|
|||||
19-040-053
|
3M™
7000127238 |
Gold-plated, Clear, Gold-plated |
Case of 10 for $3,034.50
|
|
|||||
19-040-044
|
3M™
7000127240 |
Shade 3 |
Case of 10 for $1,190.70
|
|
|||||
Description
- 3M Total Performance faceshields feature dual-coat technology that provides a durable, abrasion-resistant hard coat on the outside and an anti-fog coating on the inside. Made of durable, splash and impact-resistant polycarbonate.
- 3M provides reliable head and face protection products that meet regulated performance and safety requirements.
- 3M Catalog# 82600-00000, 82601-00000, 82602-00000, 82603-00000, 82604-00000, 82705-10000, 82706-10000
- For use with 3M H-Series Hard Hats/Headgear (H24M/7000002292, H24T/7100113761, H18/7000002291, H18-S/7000052752, H8A/7000002290, H10/7000052751, H8A-2/7000127237).
Compliance
Meets high-impact requirements of ANSI Z87.1-2010
Specifications
3M H-Series Hard Hats/Headgear | |
Polycarbonate |