Learn More
3M™ Replacement Fittings for Compressed Air Components
Replacement fitting for use with Supplied Air Hose W-9435 and W-2929.
Supplier: 3M™ 7000130790
Description
- 1/4 in. High pressure Socket
- 3M Catalog# W-3184-2 , 3M Catalog# W-3185-2
Specifications
Hose Socket | |
0.25 in. FPT twist lock steel and brass fitting | |
0.25 in. |
Air high pressure hose w-9435 and w-2929 | |
W-3185-2 | |
0.63 cm |