Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Replacement Fittings for Compressed Air Components

Replacement fitting for use with Supplied Air Hose W-9435 and W-2929.

$87.53 - $254.14

Specifications

Product Type Hose Socket
For Use With (Equipment) Air high pressure hose w-9435 and w-2929
Length (English) 0.25 in.
Length (Metric) 0.63 cm
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-048-488
View Documents
3M™
7000056515
0.25 in. FPT industrial interchange brass fitting
Case of 2 for $87.53
Only null left
Add to Cart
 
19-021-658
View Documents
3M™
7000130790
0.25 in. FPT twist lock steel and brass fitting
Case of 2 for $254.14
Only null left
Add to Cart
 
Description

Description

  • 1/4 in. High pressure Socket
  • 3M Catalog# W-3184-2 , 3M Catalog# W-3185-2
Specifications

Specifications

Hose Socket
Air high pressure hose w-9435 and w-2929
0.25 in.
0.63 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.