Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Replacement Fittings for Compressed Air Components: 0.37 Inch

Designed for use with 3M compressed air components.

$87.23 - $212.15

Specifications

Includes Twist lock steel and brass fitting
Length (English) 0.37 in.
Length (Metric) 0.93 cm
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Product Type Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Material Price Quantity & Availability  
19-813-797
View Documents
3M™
7000130788
Air high pressure hose, compressed air filter and regulator panel w-2806 Hose Socket Steel, Brass
Case of 2 for $212.15
Only null left
Add to Cart
 
19-021-567
View Documents
3M™
7100017458
Air low pressure hose w-3020 Plug Steel
Case of 2 for $87.23
Only null left
Add to Cart
 
Description

Description

Replacement parts for compressed air components
  • High pressure, 3/8 inch MPT twist lock fitting for 3M™ Compressed Air Filter and Regulator Panel W-2806
  • Low pressure, 3/8 inch FPT twist lock steel and brass fitting for 3M™ Supplied Air Hose W-3020
  • 3M Catalog# W-3182-2 , 3M Catalog# W-3198-2

Specifications

Specifications

Twist lock steel and brass fitting
0.37 in.
0.93 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.