Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Rugged Comfort 6500 Series Half Facepiece Reusable Respirator
Shop All 3M Products
Click to view available options
Size:
Small
Medium
Large
Description
- Resilient silicone faceseal offers an extended facepiece life, even keeping its shape in high-heat environments
- Sleek, low-profile design
- Special valve cover helps to direct exhaled breath and moisture downward
- 3M’s proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
- Fits securely and comfortably on the head
- Includes adjustable head harness assembly featuring long-lasting polyester/spandex straps that stretch and conform to the wearer
- Highly versatile for many workplace environments and a variety of jobs where respiratory protection is required
- Compatible with all 3M™ bayonet-style filters and cartridges, including 6000 Series Cartridges and 3M™ 2000, 2200, 7000 or 5000 Series Filters, to help provide respiratory protection against a variety of gases, vapors, and particulate hazards according to NIOSH approvals
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Certifications (OSHA) | OSHA 29 CFR Part 1920.134 |
| Connection Type | Bayonet |
| Color | Gray, Teal |
| Product Type | Half-Mask Respirator |
| Size | Medium |
| Facepiece Material | Silicone |
| For Use With (Application) | Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning |
| For Use With (Equipment) | Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters |
| Model | 6502 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction