Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Versaflo™ S-Series Headcovers and Hoods
Shop All 3M Products
Click to view available options
Product Type:
Hood with Faceseal
Hood with Inner Shroud
Hood with Neck Collar
Painter's Hood with Inner Shroud Assembly
Color:
Gray
White
Model:
S-433 L-5
S-605-10
S-607-10
S-655
S-657
S-707-10
S-757
S-805-5
Description
- S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Provides head, face, neck and shoulder coverage from dust and paint spray.
- Provides a good field of view.
- Helps protect eyes and face from liquid splash and low energy flying particles and dust.
- Versatile accessories available that can help you adapt the hood to your particular environment.

Specifications
Specifications
Assigned Protection Factor (APF) | 1000 |
Certifications/Compliance | NIOSH |
Description | Painter's Hood, S-757 |
Color | White |
Material | Polypropylene |
Product Type | Painter's Hood with Inner Shroud Assembly |
For Use With (Equipment) | 3M™ powered air purifying respirator and supplied air respirator valves |
Includes | Painter’s Hood with Inner Shroud, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off |
Material (Lens) | PETG |
Model | S-757 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction