Description
The 332AF safety impact goggles are a dependable and economical choice that offer a sturdy polycarbonate lens, adjustable elastic strap and a wide opening to fit over prescription eyewear. It is built with a direct venting system that circulates air and helps minimize fogging, allowing for use in varied environments and conditions.
- Clear lens
- Polycarbonate lens absorbs 99.9% UV
- Economical goggle
- Direct venting system that circulates air and helps minimize fogging
- Over-the-glass coverage
- Meets the High Impact Requirements of ANSI Z87.1-2010
- 10 safety goggles per case
- 3M Catalog# 40651-00000-10
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Safety Impact Goggles |
| No. per Case | 1/Cs. |
| Model | 40651-00000-10 |